siRNA Details
| Gene Symbol: Sectm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199082.2 | NP_954513.2 | 3 | 464 | ||
| XM_006247879.2 | XP_006247941.1 | 3 | 560 | |||
| XM_006247880.2 | XP_006247942.1 | 4 | 1384 | |||
| XM_006247881.2 | XP_006247943.1 | 3 | 1101 | |||
| XM_006247882.2 | XP_006247944.1 | 3 | 557 | |||
| XM_006247884.2 | XP_006247946.1 | 4 | 1382 | |||
| GenBank mRNA | BC061794.1 | - | 3 | 459 | 5597864 | |
| BQ201063.1 | - | 2 | 653 | |||
| DY309177.1 | - | 2 | 552 | |||

Sectm1b
Genomic Map