siRNA Details
| Gene Symbol: Ube2v2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183052.2 | NP_898875.1 | 3 | 234 | ||
| XM_006248727.2 | XP_006248789.1 | 3 | 268 | |||
| GenBank mRNA | AJ515244.1 | - | 3 | 222 | ||
| BC087593.1 | - | 3 | 212 | 7310168 | ||
| FM082155.1 | - | 3 | 224 | |||
| FQ216277.1 | - | 3 | 215 | |||

Ube2v2
Genomic Map