siRNA Details
| Gene Symbol: Ube2v2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183052.2 | NP_898875.1 | 4 | 405 | ||
| XM_006248727.2 | XP_006248789.1 | 4 | 439 | |||
| GenBank mRNA | BC087593.1 | - | 4 | 383 | 7310168 | |
| FM082155.1 | - | 4 | 395 | |||
| FQ216277.1 | - | 4 | 386 | |||

Ube2v2
Genomic Map