siRNA Details
| Gene Symbol: Ppm1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175755.2 | NP_786931.1 | 8 | 1164 | ||
| XM_006248674.1 | XP_006248736.1 | 8 | 1269 | |||
| XM_008768853.1 | XP_008767075.1 | 8 | 1395 | |||
| XM_008768854.1 | XP_008767076.1 | 8 | 1431 | |||
| XM_008768855.1 | XP_008767077.1 | 8 | 1449 | |||
| GenBank mRNA | AB023634.1 | - | 8 | 1164 | ||
| BC166854.1 | - | 8 | 1269 | 9098491 | ||

Ppm1f
Genomic Map