siRNA Details
| Gene Symbol: Ydjc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013863.2 | NP_001013885.2 | 5 | 667 | ||
| XM_006248682.2 | XP_006248744.1 | 5 | 836 | |||
| XM_008768859.1 | XP_008767081.1 | 6 | 700 | |||
| XM_008768860.1 | XP_008767082.1 | 7 | 1116 | |||
| XM_008768861.1 | XP_008767083.1 | 7 | 865 | |||
| GenBank mRNA | BC086565.1 | - | 5 | 817 | 7113479 | |

Ydjc
Genomic Map