siRNA Details
Gene Symbol: Psmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031639.1 | NP_001026809.1 | 9 | 1146 | ||
GenBank mRNA | BC099135.1 | - | 9 | 1146 | 7455692 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031639.1 | NP_001026809.1 | 9 | 1146 | ||
GenBank mRNA | BC099135.1 | - | 9 | 1146 | 7455692 |