siRNA Details
| Gene Symbol: Ephb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105868.1 | NP_001099338.1 | 13, 14 | 2926 | ||
| XM_006248561.2 | XP_006248623.1 | 13, 14 | 3054 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ephb3| Gene Symbol: Ephb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105868.1 | NP_001099338.1 | 13, 14 | 2926 | ||
| XM_006248561.2 | XP_006248623.1 | 13, 14 | 3054 | |||