siRNA Details
Gene Symbol: Ephb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105868.1 | NP_001099338.1 | 11 | 2465 | ||
XM_006248561.2 | XP_006248623.1 | 11 | 2593 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ephb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105868.1 | NP_001099338.1 | 11 | 2465 | ||
XM_006248561.2 | XP_006248623.1 | 11 | 2593 |