siRNA Details
| Gene Symbol: LOC102553099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248358.2 | XP_006248420.1 | 2 | 133 | ||
| Gene Symbol: Naa50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105881.1 | NP_001099351.1 | 374 | |||
| XM_006248326.2 | XP_006248388.1 | 2 | 380 | |||
| GenBank mRNA | BC169000.1 | - | 2 | 370 | 8362219 | |
| FQ216855.1 | - | 2 | 290 | |||

LOC102553099
Genomic Map