siRNA Details
| Gene Symbol: Pcnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047846.1 | NP_001041311.1 | 3 | 250 | ||
| XM_006248241.2 | XP_006248303.1 | 2 | 223 | |||
| XM_006248242.1 | XP_006248304.1 | 2 | 224 | |||
| XM_008768630.1 | XP_008766852.1 | 2 | 282 | |||
| XM_008768631.1 | XP_008766853.1 | 2 | 282 | |||
| GenBank mRNA | AY325209.1 | - | 3 | 250 | ||

Pcnp
Genomic Map