siRNA Details
| Gene Symbol: Psmg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105891.1 | NP_001099361.1 | 3, 4 | 613 | ||
| XM_006248154.2 | XP_006248216.1 | 4, 5 | 498 | |||
| XM_008768571.1 | XP_008766793.1 | 4, 5 | 770 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmg1| Gene Symbol: Psmg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105891.1 | NP_001099361.1 | 3, 4 | 613 | ||
| XM_006248154.2 | XP_006248216.1 | 4, 5 | 498 | |||
| XM_008768571.1 | XP_008766793.1 | 4, 5 | 770 | |||