siRNA Details
Gene Symbol: Psmg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105891.1 | NP_001099361.1 | 5, 6 | 948 | ||
XM_006248154.2 | XP_006248216.1 | 6, 7 | 833 | |||
XM_008768571.1 | XP_008766793.1 | 6, 7 | 1105 |
