siRNA Details
| Gene Symbol: Pop7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105918.1 | NP_001099388.1 | 2 | 398 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pop7| Gene Symbol: Pop7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105918.1 | NP_001099388.1 | 2 | 398 | ||