siRNA Details
| Gene Symbol: Ift22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011902.1 | NP_001011902.1 | 3, 4 | 304 | ||
| GenBank mRNA | BC089940.1 | - | 3, 4 | 304 | 7310111 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift22| Gene Symbol: Ift22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011902.1 | NP_001011902.1 | 3, 4 | 304 | ||
| GenBank mRNA | BC089940.1 | - | 3, 4 | 304 | 7310111 | |