siRNA Details
| Gene Symbol: Mrps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105923.1 | NP_001099393.1 | 1, 2 | 362 | ||
| XM_006249264.2 | XP_006249326.1 | 1, 2 | 82 | |||
| GenBank mRNA | FQ223326.1 | - | 1, 2 | 36 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps17| Gene Symbol: Mrps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105923.1 | NP_001099393.1 | 1, 2 | 362 | ||
| XM_006249264.2 | XP_006249326.1 | 1, 2 | 82 | |||
| GenBank mRNA | FQ223326.1 | - | 1, 2 | 36 | ||