siRNA Details
| Gene Symbol: Diablo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008292.1 | NP_001008293.1 | 6 | 721 | ||
| GenBank mRNA | BC086366.1 | - | 6 | 721 | 7305418 | |
| FQ219826.1 | - | 6 | 714 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Diablo| Gene Symbol: Diablo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008292.1 | NP_001008293.1 | 6 | 721 | ||
| GenBank mRNA | BC086366.1 | - | 6 | 721 | 7305418 | |
| FQ219826.1 | - | 6 | 714 | |||