siRNA Details
| Gene Symbol: Mtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013047.1 | NP_001013065.1 | 13 | 1473 | ||
| XM_008764483.1 | XP_008762705.1 | 12 | 1506 | |||
| XM_008764484.1 | XP_008762706.1 | 12 | 1506 | |||
| GenBank mRNA | BC079400.1 | - | 13 | 1620 | ||
| BC091225.1 | - | 13 | 1473 | 7129875 | ||
| FQ224358.1 | - | 12 | 1511 | |||

Mtm1
Genomic Map