siRNA Details
| Gene Symbol: Naca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105939.2 | NP_001099409.1 | 5 | 357 | ||
| NM_001198562.1 | NP_001185491.1 | 5 | 319 | |||
| NM_001198580.1 | NP_001185509.1 | 5 | 6097 | |||
| GenBank mRNA | BC166934.1 | - | 5 | 307 | 6922001 | |
| BP494723.1 | - | 5 | 341 | |||
| FQ221678.1 | - | 5 | 352 | |||
| FQ222263.1 | - | 5 | 292 | |||
| FQ225664.1 | - | 4 | 434 | |||
| FQ231879.1 | - | 5 | 346 | |||

Naca
Genomic Map