siRNA Details
| Gene Symbol: Trmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013870.1 | NP_001013892.1 | 8 | 952 | ||
| XM_006255236.2 | XP_006255298.1 | 9 | 3182 | |||
| XM_008772365.1 | XP_008770587.1 | 9 | 3258 | |||
| XM_008772366.1 | XP_008770588.1 | 8 | 3043 | |||
| GenBank mRNA | BC085126.1 | - | 8 | 952 | 7132178 | |

Trmt1
Genomic Map