siRNA Details
| Gene Symbol: Tcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053369.1 | NP_445821.1 | 8 | 733 | ||
| GenBank mRNA | AF149284.1 | - | 8 | 733 | ||
| FQ221525.1 | - | 487 | ||||
| FQ233045.1 | - | 4 | 422 | |||
| U09228.1 | - | 2 | 140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcf4| Gene Symbol: Tcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053369.1 | NP_445821.1 | 8 | 733 | ||
| GenBank mRNA | AF149284.1 | - | 8 | 733 | ||
| FQ221525.1 | - | 487 | ||||
| FQ233045.1 | - | 4 | 422 | |||
| U09228.1 | - | 2 | 140 | |||