siRNA Details
| Gene Symbol: Cacybp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004208.1 | NP_001004208.1 | 5 | 649 | ||
| GenBank mRNA | BC079007.1 | - | 5 | 649 | 7113129 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacybp| Gene Symbol: Cacybp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004208.1 | NP_001004208.1 | 5 | 649 | ||
| GenBank mRNA | BC079007.1 | - | 5 | 649 | 7113129 | |