siRNA Details
| Gene Symbol: Cnih4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105981.1 | NP_001099451.1 | 3, 4 | 359 | ||
| XM_008769819.1 | XP_008768041.1 | 2, 3 | 207 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnih4| Gene Symbol: Cnih4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105981.1 | NP_001099451.1 | 3, 4 | 359 | ||
| XM_008769819.1 | XP_008768041.1 | 2, 3 | 207 | |||