siRNA Details
| Gene Symbol: G0s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009632.1 | NP_001009632.1 | 2 | 99 | ||
| GenBank mRNA | BC088248.1 | - | 2 | 99 | 7369006 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

G0s2| Gene Symbol: G0s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009632.1 | NP_001009632.1 | 2 | 99 | ||
| GenBank mRNA | BC088248.1 | - | 2 | 99 | 7369006 | |