siRNA Details
| Gene Symbol: LOC100359687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750509.3 | XP_003750557.1 | 3 | 371 | ||
| Gene Symbol: Mrpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105997.2 | NP_001099467.1 | 3 | 377 | ||
| XM_008770009.1 | XP_008768231.1 | 4 | 744 | |||
| GenBank mRNA | BC168772.1 | - | 3 | 377 | 6920150 | |

LOC100359687
Genomic Map