siRNA Details
| Gene Symbol: Smtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013049.2 | NP_001013067.2 | 6 | 673 | ||
| XM_006251194.2 | XP_006251256.1 | 6 | 682 | |||
| XM_006251195.2 | XP_006251257.1 | 6 | 1156 | |||
| XM_006251196.2 | XP_006251258.1 | 6 | 682 | |||
| XM_008770259.1 | XP_008768481.1 | 6 | 657 | |||

Smtn
Genomic Map