siRNA Details
| Gene Symbol: RGD1309870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106018.1 | NP_001099488.1 | 4, 5 | 428 | ||
| XM_006251478.2 | XP_006251540.1 | 7, 8 | 1502 | |||
| XM_006251479.2 | XP_006251541.1 | 6, 7 | 1460 | |||
| XM_006251481.2 | XP_006251543.1 | 6, 7 | 703 | |||
| XM_006251482.2 | XP_006251544.1 | 6, 7 | 495 | |||

RGD1309870
Genomic Map