siRNA Details
| Gene Symbol: Upp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030025.1 | NP_001025196.1 | 4 | 388 | ||
| XM_006251483.2 | XP_006251545.1 | 5 | 757 | |||
| XM_006251484.2 | XP_006251546.1 | 5 | 663 | |||
| XM_006251485.2 | XP_006251547.1 | 5 | 607 | |||
| XM_006251487.2 | XP_006251549.1 | 5 | 754 | |||
| XM_008770270.1 | XP_008768492.1 | 5 | 581 | |||
| GenBank mRNA | BC099752.1 | - | 4 | 388 | 5623209 | |

Upp1
Genomic Map