siRNA Details
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100509.1 | NP_001093979.1 | 6 | 399 | ||
| GenBank mRNA | BC107950.1 | - | 6 | 320 | ||
| DY570284.1 | - | 6 | 399 | |||
| FQ213949.1 | - | 6 | 347 | |||
| FQ214043.1 | - | 6 | 352 | |||
| FQ227742.1 | - | 6 | 356 | |||

Psmc6
Genomic Map