siRNA Details
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100509.1 | NP_001093979.1 | 14 | 1202 | ||
| GenBank mRNA | BC107950.1 | - | 14 | 1123 | ||
| FQ213949.1 | - | 14 | 1150 | |||
| FQ214043.1 | - | 14 | 1155 | |||
| FQ227742.1 | - | 14 | 1159 | |||

Psmc6
Genomic Map