siRNA Details
| Gene Symbol: Cab39l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011917.1 | NP_001011917.1 | 9 | 999 | ||
| XM_006252098.2 | XP_006252160.1 | 10 | 1103 | |||
| XM_006252099.2 | XP_006252161.1 | 11 | 1187 | |||
| XM_006252100.2 | XP_006252162.1 | 12 | 1383 | |||
| XM_006252101.2 | XP_006252163.1 | 11 | 1299 | |||
| XM_008770742.1 | XP_008768964.1 | 11 | 1196 | |||
| XM_008770743.1 | XP_008768965.1 | 8 | 1593 | |||
| GenBank mRNA | BC083684.1 | - | 9 | 999 | 7188949 | |

Cab39l
Genomic Map