siRNA Details
| Gene Symbol: Pbk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079937.1 | NP_001073406.1 | 5, 6 | 716 | ||
| XM_006252102.1 | XP_006252164.1 | 5, 6 | 597 | |||
| XM_006252103.2 | XP_006252165.1 | 5, 6 | 575 | |||
| XM_006252104.2 | XP_006252166.1 | 5, 6 | 586 | |||
| XM_006252105.2 | XP_006252167.1 | 5, 6 | 855 | |||
| XM_006252106.2 | XP_006252168.1 | 5, 6 | 795 | |||
| GenBank mRNA | BC129099.1 | - | 5, 6 | 716 | 8375047 | |

Pbk
Genomic Map