siRNA Details
| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203340.1 | NP_976085.1 | 2 | 119 | ||
| XM_006252261.2 | XP_006252323.1 | 3 | 929 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npm2| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203340.1 | NP_976085.1 | 2 | 119 | ||
| XM_006252261.2 | XP_006252323.1 | 3 | 929 | |||