siRNA Details
| Gene Symbol: Fkbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037180.1 | NP_001032257.1 | 3 | 351 | ||
| GenBank mRNA | BC107454.1 | - | 3 | 351 | 7378812 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp8| Gene Symbol: Fkbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037180.1 | NP_001032257.1 | 3 | 351 | ||
| GenBank mRNA | BC107454.1 | - | 3 | 351 | 7378812 | |