siRNA Details
Gene Symbol: Cers1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044230.2 | NP_001037695.1 | 4 | 646 | ||
GenBank mRNA | DQ479969.1 | - | 4 | 624 |
Gene Symbol: Gdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044240.2 | NP_001037705.1 | 4 | 646 |
