siRNA Details
Gene Symbol: Ing2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106083.1 | NP_001099553.1 | 2 | 1121 | ||
XM_006253131.2 | XP_006253193.1 | 2 | 3112 | |||
XM_006253132.2 | XP_006253194.1 | 3 | 1373 | |||
XM_006253133.2 | XP_006253195.1 | 3 | 728 | |||
XM_008771255.1 | XP_008769477.1 | 2 | 992 | |||
GenBank mRNA | BC168652.1 | - | 1101 | 7097301 |
