siRNA Details
| Gene Symbol: Ing2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106083.1 | NP_001099553.1 | 2 | 1081 | ||
| XM_006253131.2 | XP_006253193.1 | 2 | 3072 | |||
| XM_006253132.2 | XP_006253194.1 | 3 | 1333 | |||
| XM_006253133.2 | XP_006253195.1 | 3 | 688 | |||
| XM_008771255.1 | XP_008769477.1 | 2 | 952 | |||
| GenBank mRNA | BC168652.1 | - | 1061 | 7097301 | ||

Ing2
Genomic Map