siRNA Details
| Gene Symbol: Uimc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013884.1 | NP_001013906.1 | 6 | 827 | ||
| XM_006253613.2 | XP_006253675.1 | 6 | 1231 | |||
| GenBank mRNA | BC087149.1 | - | 6 | 827 | 7131093 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uimc1| Gene Symbol: Uimc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013884.1 | NP_001013906.1 | 6 | 827 | ||
| XM_006253613.2 | XP_006253675.1 | 6 | 1231 | |||
| GenBank mRNA | BC087149.1 | - | 6 | 827 | 7131093 | |