siRNA Details
| Gene Symbol: Nqo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004214.1 | NP_001004214.1 | 6 | 561 | ||
| XM_006253867.2 | XP_006253929.1 | 6 | 812 | |||
| XM_006253868.2 | XP_006253930.1 | 5 | 570 | |||
| GenBank mRNA | BC079157.1 | - | 6 | 561 | 7128990 | |

Nqo2
Genomic Map