siRNA Details
| Gene Symbol: Uqcrfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008888.1 | NP_001008888.1 | 2 | 304 | ||
| GenBank mRNA | AY539946.1 | - | 2 | 249 | ||
| BC085339.1 | - | 2 | 304 | 7302533 | ||
| FQ213498.1 | - | 2 | 318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uqcrfs1| Gene Symbol: Uqcrfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008888.1 | NP_001008888.1 | 2 | 304 | ||
| GenBank mRNA | AY539946.1 | - | 2 | 249 | ||
| BC085339.1 | - | 2 | 304 | 7302533 | ||
| FQ213498.1 | - | 2 | 318 | |||