siRNA Details
| Gene Symbol: Ucma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106121.1 | NP_001099591.1 | 3 | 328 | ||
| XM_006254265.1 | XP_006254327.1 | 5 | 968 | |||
| GenBank mRNA | EU022754.1 | - | 5 | 403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ucma| Gene Symbol: Ucma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106121.1 | NP_001099591.1 | 3 | 328 | ||
| XM_006254265.1 | XP_006254327.1 | 5 | 968 | |||
| GenBank mRNA | EU022754.1 | - | 5 | 403 | ||