siRNA Details
| Gene Symbol: LOC100174910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131003.1 | NP_001124475.1 | 4 | 353 | ||
| XM_006254736.2 | XP_006254798.1 | 4 | 491 | |||
| XM_006254737.2 | XP_006254799.1 | 3 | 434 | |||
| XM_006254738.2 | XP_006254800.1 | 6 | 510 | |||
| XM_006254739.2 | XP_006254801.1 | 6 | 503 | |||
| XM_006254740.2 | XP_006254802.1 | 5 | 399 | |||
| XM_006254741.2 | XP_006254803.1 | 5 | 396 | |||
| XM_008772078.1 | XP_008770300.1 | 5 | 1866 | |||
| XM_008772079.1 | XP_008770301.1 | 4 | 1765 | |||
| GenBank mRNA | BC098717.1 | - | 3 | 927 | ||
| BC166916.1 | - | 3 | 296 | 6918206 | ||
| BC168769.1 | - | 3 | 296 | 6891274 | ||
| CB736776.1 | - | 5 | 394 | |||

LOC100174910
Genomic Map