siRNA Details
Gene Symbol: LOC100174910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131003.1 | NP_001124475.1 | 2 | 134 | ||
XM_006254736.2 | XP_006254798.1 | 2 | 272 | |||
XM_006254737.2 | XP_006254799.1 | 2 | 272 | |||
XM_006254738.2 | XP_006254800.1 | 4 | 291 | |||
XM_006254739.2 | XP_006254801.1 | 4 | 284 | |||
XM_006254740.2 | XP_006254802.1 | 3 | 180 | |||
XM_006254741.2 | XP_006254803.1 | 3 | 177 | |||
XM_008772078.1 | XP_008770300.1 | 3 | 1647 | |||
XM_008772079.1 | XP_008770301.1 | 2 | 1546 | |||
GenBank mRNA | BC098717.1 | - | 2 | 131 | ||
BC166916.1 | - | 2 | 134 | 6918206 | ||
BC168769.1 | - | 2 | 134 | 6891274 | ||
CB736776.1 | - | 3 | 175 |
