siRNA Details
| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271105.1 | NP_001258034.1 | 1 | 229 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh7a1| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271105.1 | NP_001258034.1 | 1 | 229 | ||