siRNA Details
| Gene Symbol: Ppic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004215.1 | NP_001004215.1 | 3 | 355 | ||
| GenBank mRNA | BC078949.1 | - | 3 | 355 | 7109986 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppic| Gene Symbol: Ppic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004215.1 | NP_001004215.1 | 3 | 355 | ||
| GenBank mRNA | BC078949.1 | - | 3 | 355 | 7109986 | |