siRNA Details
| Gene Symbol: Fbln5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019153.3 | NP_062026.2 | 10 | 1291 | ||
| XM_006240459.2 | XP_006240521.1 | 10 | 1522 | |||
| GenBank mRNA | AF112153.1 | - | 10 | 1415 | ||
| AF137350.1 | - | 10 | 1218 | |||
| BC078845.1 | - | 10 | 1291 | 7123752 | ||

Fbln5
Genomic Map