siRNA Details
| Gene Symbol: Tepp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201655.1 | NP_970617.1 | 4 | 583 | ||
| XM_008772274.1 | XP_008770496.1 | 4 | 860 | |||
| XM_008772275.1 | XP_008770497.1 | 4 | 860 | |||
| XM_008772276.1 | XP_008770498.1 | 4 | 860 | |||

Tepp
Genomic Map