siRNA Details
| Gene Symbol: Tepp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201655.1 | NP_970617.1 | 6, 7 | 766 | ||
| XM_008772274.1 | XP_008770496.1 | 5, 6 | 1149 | |||
| XM_008772276.1 | XP_008770498.1 | 5, 6 | 1149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tepp| Gene Symbol: Tepp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201655.1 | NP_970617.1 | 6, 7 | 766 | ||
| XM_008772274.1 | XP_008770496.1 | 5, 6 | 1149 | |||
| XM_008772276.1 | XP_008770498.1 | 5, 6 | 1149 | |||