siRNA Details
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106169.1 | NP_001099639.1 | 4, 5 | 555 | ||
| XM_006255079.2 | XP_006255141.1 | 4, 5 | 581 | |||
| XM_008772281.1 | XP_008770503.1 | 4, 5 | 950 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Drc7| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106169.1 | NP_001099639.1 | 4, 5 | 555 | ||
| XM_006255079.2 | XP_006255141.1 | 4, 5 | 581 | |||
| XM_008772281.1 | XP_008770503.1 | 4, 5 | 950 | |||