siRNA Details
| Gene Symbol: Csn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031562.1 | NP_113750.1 | 4 | 148 | ||
| GenBank mRNA | K02598.1 | - | 4 | 148 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csn3| Gene Symbol: Csn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031562.1 | NP_113750.1 | 4 | 148 | ||
| GenBank mRNA | K02598.1 | - | 4 | 148 | ||