siRNA Details
| Gene Symbol: Hsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106177.1 | NP_001099647.1 | 7 | 790 | ||
| XM_006255453.2 | XP_006255515.1 | 8 | 1344 | |||
| XM_006255454.2 | XP_006255516.1 | 8 | 1358 | |||
| XM_006255455.2 | XP_006255517.1 | 7 | 1235 | |||

Hsf4
Genomic Map